Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100359.3 | NP_001093829.1 | 8 - 9 | 1327 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |