Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100365.2 | NP_001093835.1 | 3 - 4 | 414 | 60 | |
GenBank mRNA | BC149502.1 | - | 3 - 4 | 415 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |