Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100366.1 | NP_001093836.1 | 8 - 9 | 1507 | 100 | |
GenBank mRNA | BC147910.1 | - | 8 - 9 | 1507 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |