Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101084.1 | NP_001094554.1 | 1 - 2 | 412 | 82 | |
GenBank mRNA | BC151495.1 | - | 1 - 2 | 412 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |