Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101084.1 | NP_001094554.1 | 3 - 4 | 660 | 60 | |
GenBank mRNA | BC151495.1 | - | 3 - 4 | 660 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |