Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101090.1 | NP_001094560.1 | 1 - 2 | 202 | 82 | |
GenBank mRNA | BC151577.1 | - | 1 - 2 | 202 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |