Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101098.2 | NP_001094568.1 | 3 - 4 | 702 | 63 | |
GenBank mRNA | BC151505.1 | - | 3 - 4 | 703 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |