Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101114.1 | NP_001094584.1 | 3 - 4 | 1181 | 120 | |
GenBank mRNA | BC149099.1 | - | 3 - 4 | 1181 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |