Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101852.1 | NP_001095322.1 | 1 - 2 | 386 | 61 | |
GenBank mRNA | BC150101.1 | - | 1 - 2 | 386 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |