Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101926.2 | NP_001095396.1 | 8 - 9 | 1089 | 72 | |
GenBank mRNA | BC103402.1 | - | 8 - 9 | 1116 | 72 | |
BC151320.1 | - | 8 - 9 | 1090 | 72 | ||
BT030699.1 | - | 8 - 9 | 734 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |