Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101937.1 | NP_001095407.1 | 4 - 5 | 524 | 130 | |
GenBank mRNA | BC149229.1 | - | 4 - 5 | 524 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |