Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102002.2 | NP_001095472.1 | 2 - 3 | 433 | 109 | |
GenBank mRNA | BC151468.1 | - | 2 - 3 | 434 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |