Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102002.2 | NP_001095472.1 | 4 - 5 | 708 | 78 | |
GenBank mRNA | BC151468.1 | - | 4 - 5 | 709 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |