Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102016.1 | NP_001095486.1 | 9 - 10 | 1464 | 71 | |
GenBank mRNA | BC149840.1 | - | 9 - 10 | 1464 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |