Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102042.2 | NP_001095512.2 | 11 - 12 | 1625 | 121 | |
GenBank mRNA | BC148948.1 | - | 11 - 12 | 2679 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |