Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102042.2 | NP_001095512.2 | 3 - 4 | 423 | 123 | |
GenBank mRNA | BC148948.1 | - | 3 - 4 | 423 | 123 | |
DV917263.1 | - | 2 - 3 | 238 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |