Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102506.2 | NP_001095976.1 | 3 - 4 | 2663 | 116 | |
GenBank mRNA | BC148989.1 | - | 3 - 4 | 871 | 116 | |
BC151664.1 | - | 3 - 4 | 2664 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |