Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103233.1 | 2 - 3 | 424 | 84 | ||
GenBank mRNA | BC149702.1 | - | 2 - 3 | 424 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |