Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104977.1 | NP_001098447.1 | 2 - 3 | 238 | 125 | |
GenBank mRNA | BT030712.1 | - | 2 - 3 | 238 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |