Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105331.2 | NP_001098801.2 | 1 - 2 | 120 | 139 | |
GenBank mRNA | BC153238.1 | - | 1 - 2 | 121 | 139 | |
BT026215.1 | - | 1 - 2 | 86 | 139 | ||
BT029828.1 | - | 1 - 2 | 102 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |