Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105331.2 | NP_001098801.2 | 2 - 3 | 322 | 62 | |
NM_001164088.1 | NP_001157560.1 | 2 - 3 | 337 | 62 | ||
GenBank mRNA | BC153238.1 | - | 2 - 3 | 323 | 62 | |
BT026172.1 | - | 2 - 3 | 308 | 62 | ||
BT026215.1 | - | 2 - 3 | 288 | 62 | ||
BT026330.1 | - | 2 - 3 | 327 | 62 | ||
BT029825.1 | - | 2 - 3 | 325 | 62 | ||
BT029828.1 | - | 2 - 3 | 304 | 62 | ||
DN283722.1 | - | 1 - 2 | 185 | 62 | ||
EE230608.1 | - | 2 - 3 | 338 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |