Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105356.2 | NP_001098826.1 | 4 - 5 | 423 | 65 | |
GenBank mRNA | BC134691.1 | - | 4 - 5 | 424 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |