Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109980.3 | NP_001103450.1 | 5 - 6 | 1327 | 85 | |
GenBank mRNA | AB435503.1 | - | 3 - 4 | 952 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |