Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110076.2 | NP_001103546.1 | 1 - 2 | 181 | 84 | |
GenBank mRNA | BC105414.1 | - | 1 - 2 | 182 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |