Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110078.2 | NP_001103548.2 | 3 - 4 | 341 | 82 | |
GenBank mRNA | BC122639.1 | - | 2 - 3 | 317 | 82 | |
EE338668.1 | - | 3 - 4 | 342 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |