Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113233.1 | NP_001106704.1 | 1 - 2 | 154 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |