Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113258.1 | NP_001106729.1 | 1 - 2 | 86 | 81 | |
GenBank mRNA | DV931938.1 | - | 1 - 2 | 65 | 81 | |
EE390515.1 | - | 1 - 2 | 88 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |