Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024571.2 | NP_001019742.2 | 2 - 3 | 285 | 59 | |
GenBank mRNA | BC111221.1 | - | 2 - 3 | 285 | 59 | |
BT021786.1 | - | 1 - 2 | 224 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |