Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035470.2 | NP_001030547.2 | 2 - 3 | 293 | 73 | |
GenBank mRNA | BC118296.1 | - | 2 - 3 | 293 | 73 | |
DT811968.1 | - | 2 - 3 | 248 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |