Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046422.2 | NP_001039887.2 | 4 - 5 | 709 | 114 | |
NM_001113249.1 | NP_001106720.1 | 4 - 5 | 709 | 114 | ||
GenBank mRNA | BC105361.1 | - | 4 - 5 | 692 | 114 | |
DT830273.1 | - | 4 - 5 | 495 | 114 | ||
DT837693.1 | - | 2 - 3 | 258 | 114 | ||
DT847039.1 | - | 4 - 5 | 713 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |