Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046478.2 | NP_001039943.2 | 2 - 3 | 673 | 61 | |
GenBank mRNA | DV892216.1 | - | 2 - 3 | 673 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |