Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076274.2 | NP_001069742.2 | 2 - 3 | 385 | 110 | |
GenBank mRNA | BC108166.1 | - | 2 - 3 | 201 | 110 | |
BC134728.1 | - | 2 - 3 | 385 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |