Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076426.2 | NP_001069894.2 | 1 - 2 | 86 | 125 | |
GenBank mRNA | BC102597.1 | - | 1 - 2 | 87 | 125 | |
DT841167.1 | - | 1 - 2 | 86 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |