Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076896.2 | NP_001070364.1 | 2 - 3 | 334 | 73 | |
NM_001114610.1 | NP_001108082.1 | 2 - 3 | 334 | 73 | ||
GenBank mRNA | BC109841.1 | - | 2 - 3 | 322 | 73 | |
DT811342.1 | - | 2 - 3 | 327 | 73 | ||
DY454254.1 | - | 2 - 3 | 338 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |