Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077022.2 | NP_001070490.1 | 1 - 2 | 75 | 97 | |
GenBank mRNA | BC108167.1 | - | 1 - 2 | 70 | 97 | |
BF600024.1 | - | 1 - 2 | 61 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |