Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C23H6orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077999.2 | NP_001071467.1 | 2 - 3 | 793 | 75 | |
GenBank mRNA | BC126615.1 | - | 2 - 3 | 405 | 75 | |
BC148131.1 | - | 2 - 3 | 1685 | 75 | ||
BT030562.1 | - | 2 - 3 | 793 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |