Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078150.2 | NP_001071618.1 | 3 - 4 | 1255 | 157 | |
GenBank mRNA | BC113338.1 | - | 3 - 4 | 1101 | 157 | |
BT030519.1 | - | 3 - 4 | 1255 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |