Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081743.2 | NP_001075212.1 | 4 - 5 | 705 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |