Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098059.1 | NP_001091528.1 | 8 - 9 | 1081 | 102 | |
GenBank mRNA | BT030596.2 | - | 8 - 9 | 1079 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |