Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098060.2 | NP_001091529.1 | 7 - 8 | 1382 | 81 | |
GenBank mRNA | BC140696.1 | - | 7 - 8 | 1383 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |