Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098088.2 | NP_001091557.1 | 3 - 4 | 1190 | 84 | |
GenBank mRNA | BC140541.1 | - | 3 - 4 | 1191 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |