Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098108.1 | NP_001091577.1 | 1 - 2 | 258 | 67 | |
GenBank mRNA | BC140533.1 | - | 1 - 2 | 258 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |