Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098115.1 | NP_001091584.1 | 4 - 5 | 921 | 122 | |
GenBank mRNA | BC140518.1 | - | 4 - 5 | 921 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |