Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098116.1 | NP_001091585.1 | 3 - 4 | 858 | 111 | |
GenBank mRNA | BC140469.1 | - | 3 - 4 | 858 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |