Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098123.1 | NP_001091592.1 | 4 - 5 | 2699 | 81 | |
GenBank mRNA | BC140519.1 | - | 4 - 5 | 2699 | 81 | |
BC149076.1 | - | 2 - 3 | 2223 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |