Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098385.1 | NP_001091855.1 | 2 - 3 | 770 | 89 | |
GenBank mRNA | BC134686.1 | - | 2 - 3 | 770 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |