Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099039.2 | NP_001092509.1 | 1 - 2 | 192 | 59 | |
GenBank mRNA | BC142315.1 | - | 1 - 2 | 194 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |