Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099076.1 | NP_001092546.1 | 4 - 5 | 646 | 70 | |
GenBank mRNA | BC142237.1 | - | 4 - 5 | 646 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |