Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBLAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099079.2 | NP_001092549.1 | 1 - 2 | 803 | 87 | |
GenBank mRNA | BC142228.1 | - | 1 - 2 | 804 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |