Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099096.1 | NP_001092566.1 | 1 - 2 | 464 | 91 | |
GenBank mRNA | BC146074.1 | - | 1 - 2 | 464 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |